Learn More
Alkali Scientific AlumaFoil™ Aluminum Foil PCR Sealing Film, 100/pk
SDP
Catalog No. 502389039
Encompass_Preferred
AlumaFoil™ Aluminum Foil PCR Sealing Film, PCR features a specially formulated adhesive that provides an excellent seal to minimize evaporation, reduce cross-contamination and prevent spillage. Universal fit for all plate types (e.g. flat-top, raised-rim and non-skirted); the smaller dimensions and rounded corners result in no plate overhang for robotics applications. Certified DNase- & RNase- free. Pierceable with a pipet tip or robotic probe for direct sample recovery. 2.0mil aluminum foil removes easily without shredding or tearing. The conformable nature of foil allows for the visual verification of the seal’s security around each well. Not recommended for long-term storage.
Catalog No. 50-238-9039 Supplier Alkali Scientific Supplier No. SF17ML
AlumaFoil™ Aluminum Foil PCR Sealing Film, PCR features a specially formulated adhesive that provides an excellent seal to minimize evaporation, reduce cross-contamination and prevent spillage. Universal fit for all plate types (e.g. flat-top, raised-rim and non-skirted); the smaller dimensions and rounded corners result in no plate overhang for robotics applications. Certified DNase- & RNase- free. Pierceable with a pipet tip or robotic probe for direct sample recovery. 2.0mil aluminum foil removes easily without shredding or tearing. The conformable nature of foil allows for the visual verification of the seal’s security around each well. Not recommended for long-term storage.