Learn More
Research Products International Corp Alumaseal CS Sealing Film, Non-Sterile, 100 per Package
SDP
Catalog No. 50999333
Encompass
Aluminum sealing film has a specially formulated adhesive that withstands low temperature storage to -80°C. Film is pierceable with a pipet tip for sample recovery. Features a non-perforated end tab that simplifies application. This film is not recommended for PCR applications. AlumaSeal CS is a Trademark of Excel Scientific, Inc.
Catalog No. 50-999-333 Supplier Research Products International Corp Supplier No. 202535
Aluminum sealing film has a specially formulated adhesive that withstands low temperature storage to -80°C. Film is pierceable with a pipet tip for sample recovery. Features a non-perforated end tab that simplifies application. This film is not recommended for PCR applications. AlumaSeal CS is a Trademark of Excel Scientific, Inc.