Learn More
Research Products International Corp Alumaseal CS Sealing Film, Sterile, 50 per Package
SDP
Catalog No. 50998791
Encompass
Aluminum sealing film has a specially formulated adhesive that withstands low temperature storage to -80°C. Film is pierceable with a pipet tip for sample recovery. Features a non-perforated end tab that simplifies application. This film is not recommended for PCR applications. AlumaSeal CS is a Trademark of Excel Scientific, Inc.
Catalog No. 50-998-791 Supplier Research Products International Corp Supplier No. 202536
Aluminum sealing film has a specially formulated adhesive that withstands low temperature storage to -80°C. Film is pierceable with a pipet tip for sample recovery. Features a non-perforated end tab that simplifies application. This film is not recommended for PCR applications. AlumaSeal CS is a Trademark of Excel Scientific, Inc.