Learn More
Research Products International Corp AlumaSeal II Adhesive Aluminum Sealing Foil for PCR Plates, Sterile, 50 per Package
SDP
Catalog No. 50212646
Encompass
AlumaSeal II prevents evaporation, vapor loss, cross contamination, aerosol release and spillage from multiwell PCR or assay plates.This unique sealing foil conforms around individual plate wells to provide a positive and uniform seal. Medical grade adhesive has a thermal stability range of -80°C to 120° C with superior chemical and solvent resistance. AlumaSeal ll eliminates the need for other types of plate sealing methods such as mats, caps or heat sealers. Pre-cut sheets have a 125 x 82 mm sealing area to fit both skirted and non-skirted PCR plates. Removable end tabs allow quick and accurate positioning over plate wells. Samples can be easily recovered by piercing foil with standard pipet tips or metal robotic probes. AlumaSeal II is a Trademark of Excel Scientific, Inc.
Excellent thermal stability
Piercable for sample recovery
RNase, DNase free
Catalog No. 50-212-646 Supplier Research Products International Corp Supplier No. 248898
AlumaSeal II prevents evaporation, vapor loss, cross contamination, aerosol release and spillage from multiwell PCR or assay plates.This unique sealing foil conforms around individual plate wells to provide a positive and uniform seal. Medical grade adhesive has a thermal stability range of -80°C to 120° C with superior chemical and solvent resistance. AlumaSeal ll eliminates the need for other types of plate sealing methods such as mats, caps or heat sealers. Pre-cut sheets have a 125 x 82 mm sealing area to fit both skirted and non-skirted PCR plates. Removable end tabs allow quick and accurate positioning over plate wells. Samples can be easily recovered by piercing foil with standard pipet tips or metal robotic probes. AlumaSeal II is a Trademark of Excel Scientific, Inc.
Excellent thermal stability
Piercable for sample recovery
RNase, DNase free