Learn More
Andwin Scientific ALUMASEAL II SEALING FILMS NO
SDP
Catalog No. NC0302491
Encompass_Preferred
The AlumaSeal sealing film is ideal for use with all PCR plates. The film can easily fit with plates during application and can withstand a temperature range of -80 C to +120 C. It can also be pierced with a single or multichannel pipettor and robotic probes with no difficulty. This product is free of DNase, RNase, and nucleic acids.
Catalog No. NC0302491 Supplier Andwin Scientific Supplier No. AF100
The AlumaSeal sealing film is ideal for use with all PCR plates. The film can easily fit with plates during application and can withstand a temperature range of -80 C to +120 C. It can also be pierced with a single or multichannel pipettor and robotic probes with no difficulty. This product is free of DNase, RNase, and nucleic acids.