Learn More
Andwin Scientific ALUMASEAL IITM FILMS
SDP
Catalog No. NC9487615
Encompass_Preferred
The 82.6 x 146.1 mm AlumaSeal II Aluminum Foil Sealing Film is ideal for use in sealing all PCR plates. This product removes the necessity of heat-sealing devices or mats during the process of thermal cycling. The foil is non-permeable and has a 28 m layer of strong medical-grade acrylic adhesive.
Catalog No. NC9487615 Supplier Andwin Scientific Supplier No. AFS25
The 82.6 x 146.1 mm AlumaSeal II Aluminum Foil Sealing Film is ideal for use in sealing all PCR plates. This product removes the necessity of heat-sealing devices or mats during the process of thermal cycling. The foil is non-permeable and has a 28 m layer of strong medical-grade acrylic adhesive.