Learn More
Sigma Aldrich Fine Chemicals Biosciences AlumaSealR 384 film size 3
Catalog No. 502901419
Encompass
A 38 mum thick aluminum sealing foil film with a 38 mum layer of acrylic adhesive for use with 384 well plates. AlumaSeal 384 sealing films have one end tab with no perforations.Sealing film dimensions are 137.3 mm x 82.5 mm (13.5 mm end tab)Available as non-sterile onlyPierceableRecommended temperature range between -80 C to 120 CCertified DNase- RNase- and nucleic-acid-free
Catalog No. 50-290-1419 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z721557100EA
A 38 mum thick aluminum sealing foil film with a 38 mum layer of acrylic adhesive for use with 384 well plates. AlumaSeal 384 sealing films have one end tab with no perforations.Sealing film dimensions are 137.3 mm x 82.5 mm (13.5 mm end tab)Available as non-sterile onlyPierceableRecommended temperature range between -80 C to 120 CCertified DNase- RNase- and nucleic-acid-free