Learn More
Sigma Aldrich Fine Chemicals Biosciences AlumaSealR 96 film size 38
Catalog No. 502901728
Encompass
A 38 mum thick aluminum sealing foil film with a 38 mum layer of acrylic adhesive for use with 96 well plates. fits inside the rim of raised-rim plates. AlumaSeal 96 sealing films have one partial-width end tab with no perforations.film dimensions are 127 mm x 78 mm (9.5 mm end tab)Available as non-sterile onlyPierceableRecommended temperature range between -80C to 120CCertified DNase- RNase- and nucleic-acid-free
Catalog No. 50-290-1728 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z721549100EA
A 38 mum thick aluminum sealing foil film with a 38 mum layer of acrylic adhesive for use with 96 well plates. fits inside the rim of raised-rim plates. AlumaSeal 96 sealing films have one partial-width end tab with no perforations.film dimensions are 127 mm x 78 mm (9.5 mm end tab)Available as non-sterile onlyPierceableRecommended temperature range between -80C to 120CCertified DNase- RNase- and nucleic-acid-free