Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alumina A, Activity: I, MP EcoChrom™, MP Biomedicals™

Catalog No. ICN02105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN02105 5 kg
ICN02099 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN02105 Supplier MP Biomedicals, Inc Supplier No. 0202105
Add to Cart
Add to Cart

MP EcoChrom™ Alumina A catalyses a variety of reactions that are useful industrially.

  • Alumina A is widely used to remove water from gas streams.
  • Also used for its hardness and strength.
  • It is widely used as an abrasive, including as a much less expensive substitute for industrial diamond.
  • Many types of sandpaper use aluminium oxide crystals.
  • In chemistry laboratories, alumina is a medium for chromatography, available in basic (pH 9.5), acidic (pH 4.5 when in water) and neutral formulations, as well as for alcohols, hydrocarbons, steroids, alkaloids and natural pigments.
  • Neutral alumina, (less active than the basic form) is useful for separation of aldehydes, ketones, quinones, esters, lactones, and glucosides.
  • Acidic alumina, is the weakest and least often used type.
  • Alumina is useful for separation of acid pigments and strong acids.
  • It can cause polymerization, condensation and dehydration reactions.
  • Neither acetone nor ethyl acetate should be used as eluants, the latter being subject to saponification.
  • It is used as a dosimeter for radiation protection and therapy applications for its optically stimulated luminescence properties.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 5 kg
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina A, Activity: I, Activity acc. To Brockmann: 23 l/mm
Recommended Storage Store at room temperature.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.