Learn More
Sorbent Technologies Inc Alumina for TLC, 5-25um, Acid, 1kg
SDP
Catalog No. 501138784
Encompass_Preferred
Alumina Acid for TLC has a pH of about 4.5, particle distribution of 5-25um, and a quantity of 1kg. When combined with a binding agent, this alumina can be used for the making of analytical TLC plates. It can also be used in a packed column format for extra high resolution.
Catalog No. 50-113-8784 Supplier Sorbent Technologies Inc Supplier No. 163101
Alumina Acid for TLC has a pH of about 4.5, particle distribution of 5-25um, and a quantity of 1kg. When combined with a binding agent, this alumina can be used for the making of analytical TLC plates. It can also be used in a packed column format for extra high resolution.