Learn More
Fisher Chemical™ Alumina, Acidic, Certified
Catalog No. A948500 Shop All Fisherbrand Products
Change view
Click to view available options
500g; Amber Glass
Catalog No.
A948-500 500g; Amber Glass
1 options
Catalog No. A948-500
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
  • Common adsorbent in column and thin-layer chromatography
  • Can be used for chromatographic separation of phenols, carboxylic and amino acids in an aqueous mobile phase
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 60/325 Mesh
Packaging Amber Glass
Quantity 500 g
Material Alumina