Learn More
Sorbent Technologies Inc Alumina for TLC, 5-25um, Basic, 1kg
SDP
Catalog No. 501138786
Encompass_Preferred
Alumina Basic for TLC has a pH of about 9, particle distribution of 5-25um, and a quantity of 1kg. When combined with a binding agent, this alumina can be used for the making of analytical TLC plates. It can also be used in a packed column format for extra high resolution.
Catalog No. 50-113-8786 Supplier Sorbent Technologies Inc Supplier No. 163301
Alumina Basic for TLC has a pH of about 9, particle distribution of 5-25um, and a quantity of 1kg. When combined with a binding agent, this alumina can be used for the making of analytical TLC plates. It can also be used in a packed column format for extra high resolution.