Learn More
Cole-Parmer Techne Electronic Ice Bucket Ceramic Beads
Supplier:  Cole-Parmer 2412099
Encompass_Preferred
Keep your samples cold, clean and dry with these ceramic-coated beads. The beads are resistant to chemicals, and if required they can be autoclaved at 134C. As a time saving option, the bucket of beads can be incubated overnight in the refrigerator and then simply inserted into the unit. The beads are compatible with the electronic ice bucket, model 24120-00, which is sold separately
Catalog No. 50-196-1004