Learn More
Advalue Technologies Alumina Combustion Boat
SDP
Supplier:  Advalue Technologies AL5020
Encompass
AL-5020 Alumina Combustion Boat - Capacity 20ml - Length 100mm - Width 20mm - Height 15mm
Catalog No. NC1048877