Learn More
Mti Corporation ALUMINA CRUCIBLE,40DIA X40MM H
SDP
Catalog No. NC1386292
Encompass
Alumina Crucible: High Purity 40 dia. x 40 H mm Cylindrical with Covering Lid
Catalog No. NC1386292 Supplier Mti Corporation Supplier No. CA40D40H
Alumina Crucible: High Purity 40 dia. x 40 H mm Cylindrical with Covering Lid