Learn More
Perkin Elmer US LLC Alumina Injector, 0.80 mm I.D., for Avio
Catalog No. 502113612
Encompass_Preferred
This Alumina Injector is for use with Avio 200/500 ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.
Catalog No. 50-211-3612 Supplier Perkin Elmer US LLC Supplier No. N0791180
This Alumina Injector is for use with Avio 200/500 ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.