Learn More
Perkin Elmer US LLC Alumina Injector, 1.2 mm I.D. - Entire Length of Tube for Optima 8x00
Catalog No. NC0580109
Encompass_Preferred
This Alumina Injector is for use with Optima 8x00 ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.
Catalog No. NC0580109 Supplier Perkin Elmer US LLC Supplier No. N0781007
This Alumina Injector is for use with Optima 8x00 ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.