Learn More
Perkin Elmer US LLC Alumina Injector, 1.2 mm I.D. for Optima 8x00
Catalog No. 502112273
Encompass_Preferred
This Alumina Injector is for use with Optima 8x00 ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.
Catalog No. 50-211-2273 Supplier Perkin Elmer US LLC Supplier No. N0781015
Add to Cart
Add to Cart
This Alumina Injector is for use with Optima 8x00 ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.