Learn More
Glass Expansion Alumina Injector 1.8mm
SDP
Catalog No. 501907219
Encompass_Preferred
Alumina Injector with 1.8mm ID.A 1.8mm ID Alumina Injector (48807026), designed for less volatile organic solvents and non-HF matrices. Compatible with Spectro Blue SOP Torch 50-190-7218 (31-808-3405), Spectro Blue EOP SDT 50-190-7217 (31-808-3394) and Spectro Blue TI SDT 50-190-7225 (31-808-3468).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7219 Supplier Glass Expansion Supplier No. 318083409
Alumina Injector with 1.8mm ID.A 1.8mm ID Alumina Injector (48807026), designed for less volatile organic solvents and non-HF matrices. Compatible with Spectro Blue SOP Torch 50-190-7218 (31-808-3405), Spectro Blue EOP SDT 50-190-7217 (31-808-3394) and Spectro Blue TI SDT 50-190-7225 (31-808-3468).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001