Learn More
Glass Expansion Alumina Injector 2.5mm
SDP
Catalog No. 501907215
Encompass_Preferred
Alumina Injector with 2.5mm ID.A 2.5mm ID Alumina Injector (48807027), designed for organic solvents and non-HF matrices with high total dissolved solids. Compatible with Spectro Blue SOP Torch 50-190-7218 (31-808-3405), Spectro Blue EOP SDT 50-190-7217 (31-808-3394) and Spectro Blue TI SDT 50-190-7225 (31-808-3468).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7215 Supplier Glass Expansion Supplier No. 318083390
Alumina Injector with 2.5mm ID.A 2.5mm ID Alumina Injector (48807027), designed for organic solvents and non-HF matrices with high total dissolved solids. Compatible with Spectro Blue SOP Torch 50-190-7218 (31-808-3405), Spectro Blue EOP SDT 50-190-7217 (31-808-3394) and Spectro Blue TI SDT 50-190-7225 (31-808-3468).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001