Learn More
Glass Expansion Alumina Injector for Axial D-Torch 1.8mm
SDP
Catalog No. 501907208
Encompass_Preferred
Alumina Tapered Injector with 1.8mm ID.A 1.8mm ID alumina tapered injector suitable for use with aqueous, including HF, and less volatile organic solvent-based matrices. Compatible with Agilent ICP-OES 700-ES Series Axial, Vista Axial D-Torch (501907021).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7208 Supplier Glass Expansion Supplier No. 318083325
Alumina Tapered Injector with 1.8mm ID.A 1.8mm ID alumina tapered injector suitable for use with aqueous, including HF, and less volatile organic solvent-based matrices. Compatible with Agilent ICP-OES 700-ES Series Axial, Vista Axial D-Torch (501907021).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001