Learn More
Perkin Elmer US LLC Alumina Injector, 2.0 mm I.D. for Optima 8x00
Catalog No. NC0273567
Encompass_Preferred
This Alumina Injector is for use with Optima 8x00 ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.
Catalog No. NC0273567 Supplier Perkin Elmer US LLC Supplier No. N0781014
This Alumina Injector is for use with Optima 8x00 ICP-OES series instruments. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents, e.g., xylene.