Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alumina N, 7-12μM, MP Biomedicals™

Catalog No. ICN02148 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN02148 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN02148 Supplier MP Biomedicals, Inc Supplier No. 0202148
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alumina is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 g
Content And Storage Store at room temperature.
Product Type Alumina N
Physical Form Solid
MDL Number MFCD00003424
Density 3.94g/cm3 at 20°C
CAS 1344-28-1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.