Learn More
Sorbent Technologies Inc Alumina N TLC Plates, w/UV254, polyester backed, 200um, 4x8cm, 50/pk
SDP
Catalog No. NC0515121
Encompass_Preferred
Alumina Neutral (pH 6-8) with UV254, analytical, polyester backed TLC plates. The coating thickness is 200um and dimensions are 4x8cm. There are 50 plates per pack. Applications include: terpenes, alkaloids, steroids, aliphatic and aromatic compounds. To ensure sufficient adherence an inert organic binder is added to the aqueous suspension for coating. The aluminum oxide layer has a specific surface of about 200 m2/g (BET) and a pore size of about 60A. To activate the aluminum oxide layer, we recommend heating the TLC plates for 10 minutes at 120C.
Catalog No. NC0515121 Supplier Sorbent Technologies Inc Supplier No. 0224187
Alumina Neutral (pH 6-8) with UV254, analytical, polyester backed TLC plates. The coating thickness is 200um and dimensions are 4x8cm. There are 50 plates per pack. Applications include: terpenes, alkaloids, steroids, aliphatic and aromatic compounds. To ensure sufficient adherence an inert organic binder is added to the aqueous suspension for coating. The aluminum oxide layer has a specific surface of about 200 m2/g (BET) and a pore size of about 60A. To activate the aluminum oxide layer, we recommend heating the TLC plates for 10 minutes at 120C.