Learn More
Sorbent Technologies Inc Alumina, Neutral, 32-63um, 1kg
SDP
Catalog No. 501138780
Encompass_Preferred
Alumina Neutral, is used for packing flash chromatography columns. It has a particle distribution of 32-63um and a quantity of 1kg. The approximate pH is 7.4 and specific surface area is about 200 m2/g.
Catalog No. 50-113-8780 Supplier Sorbent Technologies Inc Supplier No. 160501
Alumina Neutral, is used for packing flash chromatography columns. It has a particle distribution of 32-63um and a quantity of 1kg. The approximate pH is 7.4 and specific surface area is about 200 m2/g.