Learn More
Hoefer, Inc Notched Alumina Plates for SE250 & SE260 Mighty Small II Mini Vertical Unit
SDP
Catalog No. NC2743282
Encompass_Preferred
SE250&SE260 Vertical Electrophoresis Unit Accessory, Notched Alumina Plate, 10 x 8 cm, Quantity: 10/Pk., For Use With: SE250&SE260 Vertical Electrophoresis Unit
Catalog No. NC2743282 Supplier Hoefer, Inc Supplier No. SE202N10
SE250&SE260 Vertical Electrophoresis Unit Accessory, Notched Alumina Plate, 10 x 8 cm, Quantity: 10/Pk., For Use With: SE250&SE260 Vertical Electrophoresis Unit