Learn More
Advalue Technologies Alumina Single Bore Tube
SDP
Catalog No. NC1792739
Encompass
AL-T3N-2N3/4-36 Alumina Single bore Tube - OD 76.2mm (3") - ID 69.85mm (2.75") - Length 914mm (36")
Catalog No. NC1792739 Supplier Advalue Technologies Supplier No. ALT3N2N3/436
AL-T3N-2N3/4-36 Alumina Single bore Tube - OD 76.2mm (3") - ID 69.85mm (2.75") - Length 914mm (36")