Learn More
Electron Microscopy Sciences Alumina Slurry 0.05 µm 6 OZ
SDP
Supplier:  Electron Microscopy Sciences 5036810
Encompass_Preferred
These suspensions are water-based and contain no additives, which may interfere with polishing or contaminate the sample. All slurries are very stable and alumina is evenly dispersed. Excellent for the finest finishes.
Catalog No. 50-997-311