Learn More
Solstice Alumina SPE Columns, Alumina Neutral, 100 mg/1 mL
Catalog No. 6002720 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100
Catalog No. Quantity
600-27-20 100
1 options
Catalog No. 600-27-20 Supplier Solstice Supplier No. 9130
Add to Cart
Add to Cart
Medical-grade polypropylene with PE frits
  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100
WARNING: Reproductive Harm - www.P65Warnings.ca.gov