Learn More
Sigma Aldrich Fine Chemicals Biosciences AluminaSealTM for cold and
Catalog No. 502901053
Encompass
Ideal for cold storage moderate and high temperature incubation protection of light sensitive samples and high throughput samplesStrong 2.7 mil thick pierceable aluminum filmNon-cytotoxic and sterileTemperature stable from -80C to 110CResistant to DMSO chemicals solvents and other common reagentsTwo scored end tabs for easy positioning
Catalog No. 50-290-1053 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z740251100EA
Ideal for cold storage moderate and high temperature incubation protection of light sensitive samples and high throughput samplesStrong 2.7 mil thick pierceable aluminum filmNon-cytotoxic and sterileTemperature stable from -80C to 110CResistant to DMSO chemicals solvents and other common reagentsTwo scored end tabs for easy positioning