Learn More
Ted Pella Inc Barrier Foil Locking Pouches, 7.5 x 8.75 x 3" ID, 100/pk
SDP
Catalog No. NC0986468
Encompass
Shown with our Specimen Storage Box, this barrier bag is ideal for storage of materials sensitive to moisture and/or oxygen. The inside dimensions measure L x H x W (where W is the pouch bottom width) with a locking opening and a tear notch above the zipper. The material is aluminum foil covered multi-layered polyethylene, providing an MVTR (moisture vapor transmission rate) that is 1,000 times better than a typical poly bag. They are heat sealable with zipper style lock and resealable. Compatible with vacuum sealers. Sizes to fit all your needs. Ideal for the storage of COVID-19 test kits.
Catalog No. NC0986468 Supplier Ted Pella Inc Supplier No. 139313
Shown with our Specimen Storage Box, this barrier bag is ideal for storage of materials sensitive to moisture and/or oxygen. The inside dimensions measure L x H x W (where W is the pouch bottom width) with a locking opening and a tear notch above the zipper. The material is aluminum foil covered multi-layered polyethylene, providing an MVTR (moisture vapor transmission rate) that is 1,000 times better than a typical poly bag. They are heat sealable with zipper style lock and resealable. Compatible with vacuum sealers. Sizes to fit all your needs. Ideal for the storage of COVID-19 test kits.