Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum acetylacetonate, 97%

Catalog No. AC196971000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC196971000 100 g Plastic bottle
AC196970050 5 g Glass bottle
AC196975000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC196971000 Supplier Thermo Scientific Chemicals Supplier No. 196971000
Only null left
Add to Cart

CAS: 13963-57-0 | C15H21AlO6 | 324.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13963-57-0
Molecular Formula C15H21AlO6
Molecular Weight (g/mol) 324.31
MDL Number MFCD00000013
InChI Key KILURZWTCGSYRE-LNTINUHCSA-K
Synonym aluminum 2, 4-pentanedionate
IUPAC Name aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 190.0°C to 195.0°C
Color White to Cream
Boiling Point 315.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Complexometry)
Packaging Plastic bottle
Linear Formula (CH3COCH2COCH2)3Al
Quantity 100 g
Solubility Information Solubility in water: 2.5g/L (20°C). Other solubilities: 4.26 g/100g alcohol (20°C), 36.45 g/100g benzene (20°C), insoluble in petroleum ether
Formula Weight 324.3
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Aluminum acetylacetonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for
EINECSNumber 237-741-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.