Learn More
BURKLE INC Aluminium bottle, UN, AL 99.5, 1.28 oz. (38 ml) with cap
Catalog No. 5000025618
Encompass_Preferred
The robust and versatile aluminum bottle with tamper-evident screw cap meets the highest safety and quality standards for storing and transporting substances and retained samples.
Made of pure aluminum (AL 99.5), it is ideal for sensitive materials that must remain unaltered. Lightweight yet extremely durable, the bottles withstand high pressure and harsh conditions.
They are corrosion-resistant, protecting against rust, chemicals, and external influences. Their seamless design ensures complete emptying. The tamper-evident cap indicates opening through a broken ring.
Aluminum bottles are opaque, naturally blocking UV rays. Fully recyclable, they offer an eco-friendly packaging solution.
The uncoated interior is odorless and suitable for non-acidic, low-salt foods. UN-approved, they comply with international standards for transporting dangerous goods, ensuring maximum safety and reliability.
Catalog No. 50-000-25618 Supplier BURKLE INC Supplier No. 03270038
The robust and versatile aluminum bottle with tamper-evident screw cap meets the highest safety and quality standards for storing and transporting substances and retained samples.
Made of pure aluminum (AL 99.5), it is ideal for sensitive materials that must remain unaltered. Lightweight yet extremely durable, the bottles withstand high pressure and harsh conditions.
They are corrosion-resistant, protecting against rust, chemicals, and external influences. Their seamless design ensures complete emptying. The tamper-evident cap indicates opening through a broken ring.
Aluminum bottles are opaque, naturally blocking UV rays. Fully recyclable, they offer an eco-friendly packaging solution.
The uncoated interior is odorless and suitable for non-acidic, low-salt foods. UN-approved, they comply with international standards for transporting dangerous goods, ensuring maximum safety and reliability.