Learn More
Uline 3M 425 Aluminum Foil Tape,1 EA
Catalog No. NC0580898
Encompass
Protect parts and surfaces from chemicals, moisture, flame, heat and light. Mask surfaces for paint stripping and patch tears in truck trailers and aircraft. Seal out moisture and reflect heat in large appliances. Excellent long-term indoor and outdoor protection. Flame resistant. Meets UL 746C, UL 723, Class "L" and F.A.R. 25.853(a). Handles temperatures from -65�F to 300�F. Aluminum foil backing with acrylic adhesive.
Catalog No. NC0580898 Supplier Uline Supplier No. S10311
Protect parts and surfaces from chemicals, moisture, flame, heat and light. Mask surfaces for paint stripping and patch tears in truck trailers and aircraft. Seal out moisture and reflect heat in large appliances. Excellent long-term indoor and outdoor protection. Flame resistant. Meets UL 746C, UL 723, Class "L" and F.A.R. 25.853(a). Handles temperatures from -65�F to 300�F. Aluminum foil backing with acrylic adhesive.