Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, for chromatography, neutral, Brockmann I, 40-300 μm, 60A

Catalog No. p-3734622
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC366680010 1 kg
AC366680025 2.5 kg
AC366680250 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC366680010 Supplier Thermo Scientific Chemicals Supplier No. 366680010
Only null left
Add to Cart

Aluminum oxide, Al2O3, CAS Number-1344-28-1, abrasit, abrarex, abramax, abramant, alundum, Aluminum oxide, alpha-alumina, fasertonerde, Aluminum oxide, aloxite, 2.5kg, 2980.0 deg.C, White, 3.97g/mL, 215-691-6, 300ppm max. | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for column chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980.0°C
Melting Point 2050.0°C
Color White
pH 6.5 to 7.5 (5% aq. suspension, stirred)
Physical Form Fine Powder
Quantity 1 kg
Linear Formula Al2O3
Fieser 01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9
Merck Index 15, 350
Solubility Information Solubility in water: insoluble
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Grade Chromatography
Packaging Plastic bottle
Density 3.97g/mL
Chemical Name or Material Aluminum oxide, Activated, Neutral, Brockmann I
Show More Show Less
EINECSNumber 215-691-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.