Learn More
LabScientific Aluminum Microtube Cooling Rack
Catalog No. NC2892284
Encompass
Aluminum Cooling Block (6x7.5x1.5 ”; 15x19x3.8 cm) 96 well format;12 x 8 format for 1.5ml microtubes and 2.0 ml cryo tubes. Well depth1.4” (36mm)
Catalog No. NC2892284 Supplier LabScientific Supplier No. 3079
Aluminum Cooling Block (6x7.5x1.5 ”; 15x19x3.8 cm) 96 well format;12 x 8 format for 1.5ml microtubes and 2.0 ml cryo tubes. Well depth1.4” (36mm)