Learn More
AdipoGen Aluminon
SDP
Catalog No. 502561764
Encompass_Preferred
Chemical. CAS 569-58-4. Formula C22H14O9 . 3NH3. MW 422.3 . 51.1. Synthetic. Aluminon, the triammonium salt of aurintricarboxylic acid, is a dye commonly used to detect the presence of the aluminium ion in an aqueous solution. In addition to its use in qualitative inorganic analysis, aluminon has applications in pigment production. It forms brilliantly colored lake pigments with aluminum, chromium, iron and beryllium. Spectral data AL-Complex Absorbance at ~525nm. Aurintricarboxylic acid readily polymerizes in aqueous solution, forming a stable free radical that inhibits protein-nucleic acid interactions. It is a potent inhibitor of RNA-directed DNA polymerase and topoisomerase II. Angiogenesis inhibitor. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells.
Catalog No. 50-256-1764 Supplier AdipoGen Supplier No. CDXA0293G025
Chemical. CAS 569-58-4. Formula C22H14O9 . 3NH3. MW 422.3 . 51.1. Synthetic. Aluminon, the triammonium salt of aurintricarboxylic acid, is a dye commonly used to detect the presence of the aluminium ion in an aqueous solution. In addition to its use in qualitative inorganic analysis, aluminon has applications in pigment production. It forms brilliantly colored lake pigments with aluminum, chromium, iron and beryllium. Spectral data AL-Complex Absorbance at ~525nm. Aurintricarboxylic acid readily polymerizes in aqueous solution, forming a stable free radical that inhibits protein-nucleic acid interactions. It is a potent inhibitor of RNA-directed DNA polymerase and topoisomerase II. Angiogenesis inhibitor. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells.