Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Aluminosilicate, mesostructured

Catalog No. 502907177
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-7177 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-290-7177 Supplier Sigma Aldrich Supplier No. 64365325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Aluminosilicate, mesostructured (Al-MSU) is an analog of hexagonal MCM-41 that can be assembled by zeolite seeds. It is hydrothermally stable and strongly acidic, which makes it useful in catalysis and adsorption technology.Al-MSU can be used as an adsorbent in the formation of hybrid membranes for fuel cell applications. It can also be used as a catalyst for biomass pyrolysis and production of aromatic amine antioxidants.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 g
Linear Formula (SiO2)x(Al2O3)y
MDL Number MFCD00077645
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.