Learn More
TA Instruments Aluminum, 20 mm, Quick Change Disposable Plate, Pkg of 20, for Use with Standard Flat Plate, Requires Quick Change Disposable Plate Holder and Stainless Steel Disposable Plate Fixture, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580831 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS, Aluminum, 20 mm, Quick Change Disposable Plate, pkg of 20, is for use with P/N 402619.901 ARES-G2, APS, Standard Flat Plate. It requires the use of P/N 402751.901 ARES-G2, APS, Quick Change Disposable Plate Holder and P/N 402303.901 ARES, Stainless Steel, Disposable Plate Fixture along with P/N 403065.901 package of 10 disposable 20 mm diameter aluminum plates.
Catalog No. 50-005-80831 Supplier TA Instruments Supplier No. 402612.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS, Aluminum, 20 mm, Quick Change Disposable Plate, pkg of 20, is for use with P/N 402619.901 ARES-G2, APS, Standard Flat Plate. It requires the use of P/N 402751.901 ARES-G2, APS, Quick Change Disposable Plate Holder and P/N 402303.901 ARES, Stainless Steel, Disposable Plate Fixture along with P/N 403065.901 package of 10 disposable 20 mm diameter aluminum plates.