Learn More
TA Instruments Aluminum, 8 mm, Quick Change Disposable Plate, Pkg of 20, for Use with Standard Flat Plate, Requires Quick Change Disposable Plate Holder and Stainless Steel Disposable Plate Fixture, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580830 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS, Aluminum, 8 mm, Quick Change Disposable Plate, pkg of 20, is for use with P/N 402619.901 ARES-G2, APS, Standard Flat Plate. It requires the use of P/N 402751.901 ARES-G2, APS, Quick Change Disposable Plate Holder and P/N 402303.901 ARES, Stainless Steel, Disposable Plate Fixture along with P/N 700.01294.17 Aluminum, 8 mm Disposable Plates.
Catalog No. 50-005-80830 Supplier TA Instruments Supplier No. 402611.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS, Aluminum, 8 mm, Quick Change Disposable Plate, pkg of 20, is for use with P/N 402619.901 ARES-G2, APS, Standard Flat Plate. It requires the use of P/N 402751.901 ARES-G2, APS, Quick Change Disposable Plate Holder and P/N 402303.901 ARES, Stainless Steel, Disposable Plate Fixture along with P/N 700.01294.17 Aluminum, 8 mm Disposable Plates.