Learn More
Strem, An Ascensus Company CAS# 13963-57-0. 500g. Aluminum acetylacetonate, 98%. MFCD00000013
SDP
Catalog No. 5090114000
Encompass
CAS# 13963-57-0. 500g. Aluminum acetylacetonate, 98%. MFCD00000013. Molecular Weight: 324.31. Molecular Formula: Al(CH3COCHCOCH3)3. Color/form: white pwdr. Strem# 93-1302. http://www.strem.com/catalog/v/93-1302/
Catalog No. 50-901-14000 Supplier Strem, An Ascensus Company Supplier No. 931302500G
CAS# 13963-57-0. 500g. Aluminum acetylacetonate, 98%. MFCD00000013. Molecular Weight: 324.31. Molecular Formula: Al(CH3COCHCOCH3)3. Color/form: white pwdr. Strem# 93-1302. http://www.strem.com/catalog/v/93-1302/