Learn More
MSC Pyramex Aluminum Full Brim Adapter, Silver, Universal, 1 Ea.
Catalog No. 5000688702
Encompass
The aluminum full brim adapter is a robust mounting solution designed to integrate face protection with full brim hard hats. This accessory provides a secure platform for attaching a wide range of face shields, ensuring compatibility with most full brim style head protection.
  • Constructed from lightweight, durable aluminum alloy
  • Designed to fit most full brim hard hats
  • Accommodates a variety of face shields
  • Includes spring mount to maintain position
  • Certified to ANSI/ISEA Z87.1-2020 standards
  • Features steel hinges for reliable operation
Catalog No. 50-006-88702 Supplier MSC Supplier No. 84829100
The aluminum full brim adapter is a robust mounting solution designed to integrate face protection with full brim hard hats. This accessory provides a secure platform for attaching a wide range of face shields, ensuring compatibility with most full brim style head protection.
  • Constructed from lightweight, durable aluminum alloy
  • Designed to fit most full brim hard hats
  • Accommodates a variety of face shields
  • Includes spring mount to maintain position
  • Certified to ANSI/ISEA Z87.1-2020 standards
  • Features steel hinges for reliable operation