Learn More
Alkali Scientific SilverSeal™ Aluminum Sealing Film
SDP
Catalog No. 502336134
Encompass_Preferred
A 36 µm soft non-permeable aluminum foil sealing film with 28 µm layer of strong medical-grade acrylic adhesive, SilverSeal™ sealing films eliminate the need for heat-sealing devices or mats during thermal cycling. Each sealing film measures 82.6 x 146.1 mm and offers sufficient sealing area for all PCR plates. Length between the perforations with end tabs removed is 125.4 mm. Compared to other aluminum foils, SilverSeal™ has less tendency to roll back or itself when removing the backing paper and conforms well to the plate during application. easily pierceable with single or multichannel pipettors and robotic probes heat and cold resistant, recommended for temperatures from -80 °C to +120 °C certified DNase-, RNase-, and nucleic-acid-free excellent barrier properties, virtually no sample evaporation or drying. SF3ML is ideal for use with 96-well raised-rim and non-skirted PCR plates.
Catalog No. 50-233-6134 Supplier Alkali Scientific Supplier No. SF3ML
A 36 µm soft non-permeable aluminum foil sealing film with 28 µm layer of strong medical-grade acrylic adhesive, SilverSeal™ sealing films eliminate the need for heat-sealing devices or mats during thermal cycling. Each sealing film measures 82.6 x 146.1 mm and offers sufficient sealing area for all PCR plates. Length between the perforations with end tabs removed is 125.4 mm. Compared to other aluminum foils, SilverSeal™ has less tendency to roll back or itself when removing the backing paper and conforms well to the plate during application. easily pierceable with single or multichannel pipettors and robotic probes heat and cold resistant, recommended for temperatures from -80 °C to +120 °C certified DNase-, RNase-, and nucleic-acid-free excellent barrier properties, virtually no sample evaporation or drying. SF3ML is ideal for use with 96-well raised-rim and non-skirted PCR plates.