Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Ammonium Sulfate 12-Hydrate, Reagent Grade, Innovating Science™
SCI_ED

Catalog No. S000505 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000505 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning: Causes skin irritation, causes serious eye irritation, may cause respiratory irritation.
Catalog No. S000505 Supplier Aldon Corporation Supplier No. IS10053
Add to Cart
Add to Cart
Warning: Causes skin irritation, causes serious eye irritation, may cause respiratory irritation.

Chemical Identifiers

CAS 7784-26-1
Molecular Weight (g/mol) 453.33 g/mol

Specifications

Physical Form Crystalline Powder or Needles
Quantity 500 g
Percent Purity 100%
Grade Reagent
Packaging HDPE
Chemical Name or Material Aluminum Ammonium Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.