Learn More
Troemner™ Aluminum ASTM Class 2 Weight
Catalog No. 02218256 Shop All Troemner Products
Change view
Click to view available options
Calibration Weight; Weight: 2 mg
Catalog No.
02-218-256 Calibration Weight; Weight: 2 mg
1 options
Catalog No. 02-218-256 Supplier Troemner™ Supplier No. 80858663
Add to Cart
Add to Cart
Troemener Individual Calibration Weights are Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
  • Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Mass
Certifications/Compliance No Certificate
Material Aluminum
Case Material Polycarbonate
Weight (Metric) 2 mg
Shape Flat
Includes Protective Case