Learn More
Research Products International Corp Aluminum Block Holds 6x50ml Tubes
SDP
Catalog No. 501977253
Encompass
Aluminum block with 6 holes for holding 50 ml centrifuge tubes in chilling and heating dry baths.
Catalog No. 50-197-7253 Supplier Research Products International Corp Supplier No. 248887
Aluminum block with 6 holes for holding 50 ml centrifuge tubes in chilling and heating dry baths.