Learn More
Strem, An Ascensus Company CAS# 7727-15-3. 50g. Aluminum bromide, 99%. MFCD00003421
SDP
Supplier:  Strem, An Ascensus Company 93138550G
Encompass
CAS# 7727-15-3. 50g. Aluminum bromide, 99%. MFCD00003421. Molecular Weight: 266.71. Molecular Formula: AlBr3. Color/form: white to off-white lumps Strem# 93-1385. http://www.strem.com/catalog/v/93-1385/
Catalog No. 50-901-14061
 Restricted Item