Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum bromide, ultra dry, 99.999% (metals basis)

Catalog No. AA4716806
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4716806 5 g
AA4716809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4716806 Supplier Thermo Scientific Chemicals Supplier No. 04716806
Add to Cart

Used as a catalyst. | CAS: 7727-15-3 | AlBr3 | 266.69 g/mol

It is used as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-15-3
Molecular Formula AlBr3
Molecular Weight (g/mol) 266.69
MDL Number MFCD00003421
InChI Key PQLAYKMGZDUDLQ-UHFFFAOYSA-K
IUPAC Name aluminium(3+) tribromide
SMILES [Al+3].[Br-].[Br-].[Br-]

Specifications

Boiling Point 265°C
Melting Point 97.5°C
Physical Form Powder
Quantity 5 g
UN Number UN1725
Solubility Information Soluble in benzene,nitrobenzene,toluene,xylene,ether,simple hydrocarbons,alcohol,carbon disulfide,ether.
Formula Weight 266.71
Percent Purity 99.999%
Packaging Plastic bottle; Ampouled under argon
Sensitivity Moisture sensitive
Density 3.21
Chemical Name or Material Aluminum bromide
Show More Show Less
Hazard Category H302-H314
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 231-779-7
RTECSNumber BD0350000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.