Learn More
Aluminum bromide, ultra dry, 99.999% (metals basis)
Description
It is used as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 265°C |
| Melting Point | 97.5°C |
| Physical Form | Powder |
| Quantity | 10 g |
| Packaging | Plastic bottle; Ampouled under argon |
| Sensitivity | Moisture sensitive |
| Density | 3.21 |
| UN Number | UN1725 |
| Solubility Information | Soluble in benzene,nitrobenzene,toluene,xylene,ether,simple hydrocarbons,alcohol,carbon disulfide,ether. |
| Formula Weight | 266.71 |
| Show More |
Safety and Handling
| Hazard Category | H302-H314 |
| Precautionary Statement | P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 231-779-7 |
| RTECSNumber | BD0350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.