Learn More
Alkali Scientific Aluminum Canisters for Blood Bag # 4R 2110
SDP
Catalog No. 5000663073
Encompass_Preferred
Canisters are constructed of light aluminum and are designed for most popular brands of of Storage Bags, or use our Universal line of Storage Canisters. All of Canisters have both a locking handle and thumb hole and can be autoclaved. Stainless Steel Canister Frames are available for all Canisters and offer a range of heights to maximize your freezer canister storage space. Description: BAXTER/FENWAL Blood Bag Canister Blood Bag Number 4R 2110 Dimensions L x D x H 10 x 8 1/16 x 13/16″ 254 x 204 x 20mm Frames Available F C 4R2110 3 F C 4R2110 4
Catalog No. 50-006-63073 Supplier Alkali Scientific Supplier No. C4R2110
Canisters are constructed of light aluminum and are designed for most popular brands of of Storage Bags, or use our Universal line of Storage Canisters. All of Canisters have both a locking handle and thumb hole and can be autoclaved. Stainless Steel Canister Frames are available for all Canisters and offer a range of heights to maximize your freezer canister storage space. Description: BAXTER/FENWAL Blood Bag Canister Blood Bag Number 4R 2110 Dimensions L x D x H 10 x 8 1/16 x 13/16″ 254 x 204 x 20mm Frames Available F C 4R2110 3 F C 4R2110 4